missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-nitroanisole, 99%

Product Code. 11386619
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11386619

Brand: Thermo Scientific Alfa Aesar H26204.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14804-39-8 | C9H11NO3 | 181.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14804-39-8
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00024537
InChI Key HSDNHFOJTRMGER-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-nitroanisole, m-xylene, 2-methoxy-5-nitro, benzene,2-methoxy-1,3-dimethyl-5-nitro, acmc-20ao84, m-xylene, 2-methoxy-5-nitro-,, 4-methoxy-3,5-dimethylnitrobenzene, 2,6-dimethyl-1-methoxy-4-nitrobenzene, 2-methoxy-1,3-di-methyl-5-nitrobenzene, benzene, 2-methoxy-1,3-dimethyl-5-nitro
PubChem CID 139835
IUPAC Name 2-methoxy-1,3-dimethyl-5-nitrobenzene
SMILES COC1=C(C)C=C(C=C1C)[N+]([O-])=O

Specifications

Melting Point 85°C to 92°C
Quantity 1 g
Formula Weight 181.19
Percent Purity 99%
Chemical Name or Material 2,6-Dimethyl-4-nitroanisole
compliance-icons
Product Identifier
  • 2,6-Dimethyl-4-nitroanisole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,6-Dimethyl-4-nitroanisole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.