missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-heptanone, technical, remainder mainly 4,6-dimethyl-2-heptanone

Artikelnummer. 10736361 Alle ansehen Thermo Scientific Chemicals Produkte
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Metal drum
missing translation for 'unitSize'
1 L
2.5 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging unitSize
10736361 20 L Metal drum 20 L
10645582 1 L Glass Bottle 1 L
10081610 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10736361 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 149440200
 Restricted Item
Estimated Shipment: 17-09-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Formats
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 108-83-8 | C9H18O | 142.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 108-83-8 , 19549-80-5
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00008940
InChI Key PTTPXKJBFFKCEK-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-heptanone, diisobutyl ketone, isovalerone, isobutyl ketone, valerone, diisobutylketone, diisobutilchetone, s-diisopropylacetone, di-isobutylcetone, dibk
PubChem CID 7958
IUPAC Name 2,6-dimethylheptan-4-one
SMILES CC(C)CC(=O)CC(C)C

Spezifikation

CAS Min % 75.0
CAS Max % 100.0
Melting Point -46°C
Color Colorless to Yellow
Density 0.8090g/mL
Boiling Point 169°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 80%
Packaging Metal drum
Linear Formula (CH3)2CHCH2COCH2CH(CH3)2
Refractive Index 1.4120 to 1.415
Quantity 20 L
Beilstein 01, 710
Specific Gravity 0.809
Solubility Information Solubility in water: slightly soluble (0.05g/100 mL). Other solubilities: miscible with most organic solvents
Viscosity 1.05 mPa.s (20°C)
Formula Weight 142.24
Percent Purity ≥75% (typically) exact percentage on the certificate of analysis, 96% min. (main compound plus isomer)
Grade Technical
Physical Form Liquid
Chemical Name or Material 2, 6-Dimethyl-4-heptanone, tech., 80%
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • 2,6-Dimethyl-4-heptanone, remainder mainly 4,6-dimethyl-2-heptanone
Signalwort
  • Danger
Gefahrenkategorie
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.