missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethyl-4-heptanone, technical, remainder mainly 4,6-dimethyl-2-heptanone, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Metal drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 75.0 |
CAS Max % | 100.0 |
Melting Point | -46°C |
Color | Colorless to Yellow |
Density | 0.8090g/mL |
Boiling Point | 169°C |
Flash Point | 49°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 80% |
Packaging | Glass Bottle |
Show More |
Safety and Handling
Product Identifier
- 2,6-Dimethyl-4-heptanone, remainder mainly 4,6-dimethyl-2-heptanone
- Danger
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'