missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-heptanone, technical, remainder mainly 4,6-dimethyl-2-heptanone

Product Code. p-118117 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
2.5 L
20 L
Product Code. Quantity Packaging unitSize
10645582 1 L Glass Bottle 1 L
10081610 2.5 L Glass Bottle 2.5 L
10736361 20 L Metal drum 20 L
3 options
This item is not returnable. View return policy

Product Code. 10645582

Brand: Thermo Scientific Acros 149440010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-83-8 | C9H18O | 142.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-83-8 , 19549-80-5
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00008940
InChI Key PTTPXKJBFFKCEK-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-heptanone, diisobutyl ketone, isovalerone, isobutyl ketone, valerone, diisobutylketone, diisobutilchetone, s-diisopropylacetone, di-isobutylcetone, dibk
PubChem CID 7958
IUPAC Name 2,6-dimethylheptan-4-one
SMILES CC(C)CC(=O)CC(C)C

Specifications

CAS Min % 75.0
CAS Max % 100.0
Melting Point -46°C
Color Colorless to Yellow
Density 0.8090g/mL
Boiling Point 169°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 80%
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2COCH2CH(CH3)2
Refractive Index 1.4120 to 1.415
Quantity 1 L
Beilstein 01, 710
Specific Gravity 0.809
Solubility Information Solubility in water: slightly soluble (0.05g/100 mL). Other solubilities: miscible with most organic solvents
Viscosity 1.05 mPa.s (20°C)
Formula Weight 142.24
Percent Purity ≥75% (typically) exact percentage on the certificate of analysis, 96% min. (main compound plus isomer)
Grade Technical
Physical Form Liquid
Chemical Name or Material 2, 6-Dimethyl-4-heptanone, tech., 80%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,6-Dimethyl-4-heptanone, remainder mainly 4,6-dimethyl-2-heptanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.