missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-3,5-heptanedione, 99%, Thermo Scientific Chemicals

Product Code. 10140572
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18362-64-6
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00015040
InChI Key CEGGECULKVTYMM-UHFFFAOYSA-N
Synonym 2,6-dimethyl-3,5-heptanedione, diisobutyrylmethane, 3,5-heptanedione, 2,6-dimethyl, acmc-209ekw, 2,6-dimethyl-3,5-heptandione
PubChem CID 87597
IUPAC Name 2,6-dimethylheptane-3,5-dione
SMILES CC(C)C(=O)CC(=O)C(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4586g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula (CH3)2CHCOCH2COCH(CH3)2
Refractive Index 1.4560 to 1.4600 (20°C, 589nm)
Quantity 5 g
Formula Weight 156.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 6-Dimethyl-3, 5-heptanedione, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,6-Dimethyl-3,5-heptanedione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'