missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-3,5-heptanedione, 97+%

Product Code. 11395017
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11395017 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11395017 Supplier Thermo Scientific Alfa Aesar Supplier No. 042005.06
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Metal chelating agent; preparation of metal diketonates for MOCVD | CAS: 18362-64-6 | C9H16O2 | 156.225 g/mol

2,6-Dimethyl-3,5-heptanedione is a metal chelating agent and it is used in the preparation of metal diketonates for Metal-Organic Chemical Vapor Deposition (MOCVD).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18362-64-6
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00015040
InChI Key CEGGECULKVTYMM-UHFFFAOYSA-N
Synonym 2,6-dimethyl-3,5-heptanedione, diisobutyrylmethane, 3,5-heptanedione, 2,6-dimethyl, acmc-209ekw, 2,6-dimethyl-3,5-heptandione
PubChem CID 87597
IUPAC Name 2,6-dimethylheptane-3,5-dione
SMILES CC(C)C(=O)CC(=O)C(C)C

Specifications

Density 0.91 g/mL
Quantity 5 g
UN Number UN1224
Solubility Information Slightly Soluble in water. (3.9g/L) (25°C)
Formula Weight 156.22
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2,6-Dimethyl-3,5-heptanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.