missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-1,4-dioxane (>85%), TRC

Product Code. 30387560
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387560 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387560 Supplier Toronto Research Chemicals Supplier No. TRCD4798855MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,6-Dimethyl-1,4-dioxane
CAS 10138-17-7
Molecular Formula C6 H12 O2
Quantity 5 mg
Synonym 2,6-Dimethyl-1,4-Dioxane
InChI Formula InChI=1S/C6H12O2/c1-5-3-7-4-6(2)8-5/h5-6H,3-4H2,1-2H3
SMILES CC1COCC(C)O1
IUPAC Name 2,6-dimethyl-1,4-dioxane
Molecular Weight (g/mol) 116.158
Formula Weight 116.084
Product Title
Select an issue

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.