missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxypyridine-3-carboxylic acid, 99%

Product Code. 10744872 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 10744872

Brand: Thermo Scientific Acros 436650100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16727-43-8 | C8H9NO4 | 183.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 16727-43-8
CAS Min % 98.5
CAS Max % 100.0
Melting Point 141.5°C to 142.8°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H9NO4
Quantity 10 g
Synonym 2,6-dimethoxynicotinic acid, 3-pyridinecarboxylic acid, 2,6-dimethoxy, 2,6-dimethoxy-nicotinic acid, pubchem11091, acmc-1bphc, 3-pyridinecarboxylicacid, 2,6-dimethoxy, 2,6-dimethoxypyridine-3-carboxylic acid, 2,6-dimethoxy-3-pyridinecarboxylic acid
InChI Key SDJQZCSCHUIITF-UHFFFAOYSA-N
SMILES COC1=NC(=C(C=C1)C(=O)O)OC
IUPAC Name 2,6-dimethoxypyridine-3-carboxylic acid
Molecular Weight (g/mol) 183.17
PubChem CID 853178
Formula Weight 183.17
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Dimethoxypyridine-3-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dimethoxypyridine-3-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.