missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxypyridine-3-boronic acid, 95%

Product Code. 11375659
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11375659 5 g 5g
11355659 250 mg 250mg
11365659 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375659 Supplier Thermo Scientific Alfa Aesar Supplier No. L20389.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 221006-70-8 | C7H10BNO4 | 182.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 221006-70-8
Molecular Formula C7H10BNO4
Molecular Weight (g/mol) 182.97
MDL Number MFCD03788239
InChI Key ADGHSWFUZUADDH-UHFFFAOYSA-N
Synonym 2,6-dimethoxypyridine-3-boronic acid, 2,6-dimethoxy-3-pyridineboronic acid, 2,6-dimethoxypyridin-3-yl boronic acid, 2,6-dimethoxypyridin-3-yl-3-boronic acid, 3-borono-2,6-dimethoxypyridine, 2,6-dimethoxy-3-pyridylboronic acid, 2,6-dimethoxypyridine-5-boronic acid, boronic acid, 2,6-dimethoxy-3-pyridinyl, 2,6-dimethoxy-3-pyridyl boronic acid
PubChem CID 2762707
IUPAC Name (2,6-dimethoxypyridin-3-yl)boronic acid
SMILES B(C1=C(N=C(C=C1)OC)OC)(O)O

Specifications

Melting Point 128°C to 130°C
Quantity 5 g
Beilstein 9391039
Formula Weight 182.97
Percent Purity 95%
Chemical Name or Material 2,6-Dimethoxypyridine-3-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.