missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxybenzonitrile, 97%, Thermo Scientific™

Product Code. 10626012 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10626012 10 g Glass bottle 10g
10539263 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10626012 Supplier Thermo Scientific Acros Supplier No. 181720100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16932-49-3
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00001788
InChI Key XHAHKSSLDJIEDH-UHFFFAOYSA-N
Synonym benzonitrile, 2,6-dimethoxy, 2,6-dimethoxy-benzonitrile, 2,6-dimethoxybenzenecarbonitrile, pubchem4772, 2.6-dimethoxybenzonitrile, benzonitrile,2,6-dimethoxy, 2-10-00-00260 beilstein handbook reference, 2,6-dimethoxybenzonitrile
PubChem CID 85648
IUPAC Name 2,6-dimethoxybenzonitrile
SMILES COC1=C(C(=CC=C1)OC)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 119°C to 123°C
Color Beige
Boiling Point 310°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CN
Quantity 10 g
Beilstein 10, 388
Formula Weight 163.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Dimethoxybenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dimethoxybenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.