missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxybenzoic acid, 99%

Product Code. 11467047
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467047 25 g 25g
11477047 100 g 100g
11484050 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467047 Supplier Thermo Scientific Alfa Aesar Supplier No. A14418.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1466-76-8 | C9H10O4 | 182.18 g/mol

It is used in the synthesis of indoles as well as coumarins related to Novobiocin, used as potent inhibitors of heat-shock protein 90 in the treatment of breast cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of indoles as well as coumarins related to Novobiocin, used as potent inhibitors of heat-shock protein 90 in the treatment of breast cancer.

Solubility
Insoluble in water. Solubility in methanol is almost transparent. Soluble in DMSO, Ethyl Acetate and acetone.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1466-76-8
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002437
InChI Key MBIZFBDREVRUHY-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-dimethoxy, 2,6-dimethoxy-benzoic acid, unii-g6b3p1lo43, 2,6-dimethoxybenzoicacid, pubchem19743, acmc-1bwjj, intermediates-zcf02125, benzoic acid,6-dimethoxy, dsstox_cid_26999, dsstox_rid_82043
PubChem CID 15109
IUPAC Name 2,6-dimethoxybenzoic acid
SMILES COC1=CC=CC(OC)=C1C(O)=O

Specifications

Melting Point ∼188°C (decomposition)
Odor Odorless
Quantity 25 g
Beilstein 2212190
Solubility Information Insoluble in water. Solubility in methanol is almost transparent. Soluble in DMSO,Ethyl Acetate and acetone.
Formula Weight 182.18
Percent Purity 99%
Chemical Name or Material 2,6-Dimethoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.