missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dimethoxyacetophenone, 98%

Product Code. 11462738
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462738 25 g 25g
11452738 5 g 5g
11472738 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462738 Supplier Thermo Scientific Alfa Aesar Supplier No. A16733.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2040-04-2 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2040-04-2
MDL Number MFCD00008729

Specifications

Melting Point 68°C to 71°C
Boiling Point 135°C to 136°C (2 mmHg)
Molecular Formula C10H12O3
Quantity 25 g
Beilstein 2048976
Synonym 2',6'-dimethoxyacetophenone, 1-2,6-dimethoxyphenyl ethanone, 2,6-dimethoxyacetophenone, acetophenone, 2',6'-dimethoxy, 1-2,6-dimethoxyphenyl ethan-1-one, ethanone, 1-2,6-dimethoxyphenyl, usaf k-2801, 2,6-dimethoxy acetophenone, 1-acetyl-2,6-dimethoxybenzene, pubchem3380
InChI Key XEUGKOFTNAYMMX-UHFFFAOYSA-N
SMILES CC(=O)C1=C(C=CC=C1OC)OC
IUPAC Name 1-(2,6-dimethoxyphenyl)ethanone
Molecular Weight (g/mol) 180.20
PubChem CID 16267
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material 2',6'-Dimethoxyacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.