missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropylphenyl isocyanate, 94%

Product Code. 11382896
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382896 25 g 25g
11362896 1 g 1g
11372896 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11382896 Supplier Thermo Scientific Alfa Aesar Supplier No. L10613.14
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 28178-42-9 | C13H17NO | 203.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28178-42-9
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.29
MDL Number MFCD00008882
InChI Key FEUFNKALUGDEMQ-UHFFFAOYSA-N
Synonym 2,6-diisopropylphenyl isocyanate, 2,6-diisopropylphenylisocyanate, 2-isocyanato-1,3-diisopropylbenzene, benzene, 2-isocyanato-1,3-bis 1-methylethyl, isocyanic acid 2,6-diisopropylphenyl ester, isocyanic acid, 2,6-diisopropylphenyl ester, 1,3-bis 1-methylethyl-2-isocyanatobenzene, 2,6-bis 1-methylethyl phenyl isocyanate, 2-isocyanato-1,3-di propan-2-yl benzene, benzene, 1,3-bis 1-methylethyl-2-isocyanato
PubChem CID 92259
IUPAC Name 2-isocyanato-1,3-di(propan-2-yl)benzene
SMILES CC(C)C1=CC=CC(C(C)C)=C1N=C=O

Specifications

Density 0.951
Boiling Point 116°C to 117°C (10 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.518
Quantity 25 g
UN Number UN2206
Beilstein 2645721
Sensitivity Moisture sensitive
Formula Weight 203.29
Percent Purity 94%
Chemical Name or Material 2,6-Diisopropylphenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.