missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Diisopropylphenol, 97%, Thermo Scientific™
Click to view available options
Quantity:
25g
100g
500g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2,6-Diisopropylphenol plays the role of an antioxidant by reacting with free radicals to form a phenoxyl radical, a property associated with all phenol-based free radical scavengers. It is an effective intravenous hypnotic agent, widely used for the induction and maintenance of anesthesia and for sedation in the intensive care unit. It is useful for the induction and maintenance of general anaesthesia.
Solubility
Very slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
2,6-Diisopropylphenol plays the role of an antioxidant by reacting with free radicals to form a phenoxyl radical, a property associated with all phenol-based free radical scavengers. It is an effective intravenous hypnotic agent, widely used for the induction and maintenance of anesthesia and for sedation in the intensive care unit. It is useful for the induction and maintenance of general anaesthesia.
Solubility
Very slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
Density | 0.962g/mL |
Boiling Point | 254°C to 256°C |
Refractive Index | 1.5140 |
Quantity | 500g |
Beilstein | 1866484 |
Merck Index | 14,7834 |
Formula Weight | 178.28 |
Percent Purity | 97% |
Chemical Name or Material | 2,6-Diisopropylphenol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'