missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropylaniline, 90+%

Product Code. 11322986
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322986 500 g 500g
11312986 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11322986 Supplier Thermo Scientific Alfa Aesar Supplier No. L10761.36
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 24544-04-5 | C12H19N | 177.291 g/mol

2,6-Diisopropylaniline acts as an intermediate used in the production of carbodiimides stabilizers, synthetic resins, antioxidants and active pharmaceutical ingredients. It is also used in the preparation of multitopic Schiff-base ligand precursors and 4,5-bis(2,6-diisopropylanilino)-2,7-di-tert-butyl-9,9-dimethylthioxanthene. It also undergoes condensation reaction with triacetylmethane to get -[1-(2,6-diisopropylphenylamino)ethylidene]pentane-2,4-dione. Further, it is used to prepare N-heterocyclic carbene complexes for alfa-arylation of acyclic ketones, amination of haloarenes and aqueous Suzuki coupling. In addition to this, it is used in the prepartion of organocatalyst based on naphthalene diimides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diisopropylaniline acts as an intermediate used in the production of carbodiimides stabilizers, synthetic resins, antioxidants and active pharmaceutical ingredients. It is also used in the preparation of multitopic Schiff-base ligand precursors and 4,5-bis(2,6-diisopropylanilino)-2,7-di-tert-butyl-9,9-dimethylthioxanthene. It also undergoes condensation reaction with triacetylmethane to get -[1-(2,6-diisopropylphenylamino)ethylidene]pentane-2,4-dione. Further, it is used to prepare N-heterocyclic carbene complexes for alfa-arylation of acyclic ketones, amination of haloarenes and aqueous Suzuki coupling. In addition to this, it is used in the prepartion of organocatalyst based on naphthalene diimides.

Solubility
Miscible with ether and alcohol. Slightly miscible with water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24544-04-5
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
MDL Number MFCD00008887
InChI Key WKBALTUBRZPIPZ-UHFFFAOYSA-N
Synonym 2,6-diisopropylaniline, benzenamine, 2,6-bis 1-methylethyl, 2,6-diisopropyl aniline, aniline, 2,6-diisopropyl, diisopropylaniline, 2,6-diisopropylphenylamine, 2,6-bis propan-2-yl aniline, 2,6-dilsopropylaniline, 2,6-di propan-2-yl aniline, 2,6-bis 1-methylethyl aniline
PubChem CID 32484
IUPAC Name 2,6-di(propan-2-yl)aniline
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N

Specifications

Melting Point -54°C
Density 0.94
Boiling Point 254°C to 257°C
Flash Point 125°C (257°F)
Refractive Index 1.533
Quantity 500 g
Beilstein 2208763
Solubility Information Miscible with ether and alcohol. Slightly miscible with water.
Formula Weight 177.29
Percent Purity ≥90%
Chemical Name or Material 2,6-Diisopropylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.