missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropyl-N,N-dimethylaniline, 96%

Product Code. 11322319
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322319 1 g 1g
11332319 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322319 Supplier Thermo Scientific Alfa Aesar Supplier No. H27275.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2909-77-5 | C14H23N | 205.345 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2909-77-5
Molecular Formula C14H23N
Molecular Weight (g/mol) 205.345
MDL Number MFCD00144978
InChI Key ALXIOUGHHXXLKX-UHFFFAOYSA-N
Synonym 2,6-diisopropyl-n,n-dimethylaniline, n,n-dimethyl-2,6-diisopropylaniline, benzenamine, n,n-dimethyl-2,6-bis 1-methylethyl, 2,6-diisopropyl-n,n-dimethyl-aniline, acmc-20ao8g, 2,6-bis methylethyl phenyl dimethylamine, n,n-dimethyl-2,6-di propan-2-yl aniline, benzenamine,n,n-dimethyl-2,6-bis 1-methylethyl
PubChem CID 76198
IUPAC Name N,N-dimethyl-2,6-di(propan-2-yl)aniline
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N(C)C

Specifications

Density 0.873
Boiling Point 86°C (2.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5005
Quantity 1 g
UN Number UN2810
Formula Weight 205.34
Percent Purity 96%
Chemical Name or Material 2,6-Diisopropyl-N,N-dimethylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.