missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dihydroxynaphthalene, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2,6-Dihydroxynaphthalene |
| CAS | 581-43-1 |
| Molecular Formula | C10H8O2 |
| Quantity | 10 g |
| Synonym | 2,6-Naphthalenediol, 2,6-Dihydroxynaphthaline, 2,6-Naphthohydroquinone, 2-Hydroxy-6-naphthol, 6-Hydroxy-2-naphthol, C.I. 76640, NSC 62687 |
| InChI Formula | InChI=1S/C10H8O2/c11-9-3-1-7-5-10(12)4-2-8(7)6-9/h1-6,11-12H |
| SMILES | Oc1ccc2cc(O)ccc2c1 |
| Molecular Weight (g/mol) | 160.17 |
| Formula Weight | 160.0524 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction