missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dihydroxynaphthalene, 98%

Product Code. 11496984
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11496984

Brand: Thermo Scientific Alfa Aesar B21465.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 581-43-1 | C10H8O2 | 160.172 g/mol

2,6-Dihydroxynaphthalene is used in the preparation of 1,5-dichloro-2,6-diethynylnaphthalenes. It is used to produce convulsions in mice. It exhibits antiproliferative activity towards certain cells. It is also employed in the preparation of the first-generation rotaxane dendrimer. Further, it is used in the production of several dyes, pigments, fluorescent whiteners, tanning agents, antioxidants and antiseptics. In addition to this, it is converted into amines, esters, ethers and carboxylic derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dihydroxynaphthalene is used in the preparation of 1,5-dichloro-2,6-diethynylnaphthalenes. It is used to produce convulsions in mice. It exhibits antiproliferative activity towards certain cells. It is also employed in the preparation of the first-generation rotaxane dendrimer. Further, it is used in the production of several dyes, pigments, fluorescent whiteners, tanning agents, antioxidants and antiseptics. In addition to this, it is converted into amines, esters, ethers and carboxylic derivatives.

Solubility
Soluble in methanol and hot water. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 581-43-1
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00004082
InChI Key MNZMMCVIXORAQL-UHFFFAOYSA-N
Synonym 2,6-dihydroxynaphthalene, 2,6-naphthalenediol, 2,6-naphthohydroquinone, 2-hydroxy-6-naphthol, unii-4xx2nd0257, 2,6-dihydroxy naphthalene, 2,6-dihdroxynaphthalene, 2,6-dihydroxy-naphthalene, acmc-209m2x, aronis24267
PubChem CID 93552
IUPAC Name naphthalene-2,6-diol
SMILES C1=CC2=C(C=CC(=C2)O)C=C1O

Specifications

Melting Point 223°C to 225°C
Quantity 100 g
Beilstein 1238082
Solubility Information Soluble in methanol and hot water. Slightly soluble in water.
Formula Weight 160.17
Percent Purity 98%
Chemical Name or Material 2,6-Dihydroxynaphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.