missing translation for 'onlineSavingsMsg'
Learn More

2,6-dihydroxyisonicotinic acid, Thermo Scientific™

Product Code. 10390593
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10390593 50 g 50g
10407164 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10390593 Supplier Thermo Scientific Maybridge Supplier No. SB00524EE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 99-11-6
Molecular Formula C6H5NO4
Molecular Weight (g/mol) 155.11
MDL Number MFCD00006274
InChI Key CSGQJHQYWJLPKY-UHFFFAOYSA-N
Synonym citrazinic acid, 2,6-dihydroxyisonicotinic acid, 2,6-dihydroxypyridine-4-carboxylic acid, 2,6-dihydroxy-4-carboxypyridine, 2,6-dihydroxy-4-pyridinecarboxylic acid, kyselina citrazinova, 2,6-dihydroxyisonicotinate, 4-pyridinecarboxylic acid, 2,6-dihydroxy, citrazinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-6-hydroxy-2-oxo
PubChem CID 7425
IUPAC Name 2-hydroxy-6-oxo-1H-pyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(=O)NC(O)=C1

Specifications

Assay Percent Range 90%
Linear Formula (HO)2(C5H2N)COOH
Quantity 50 g
Formula Weight 155.11
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2, 6-Dihydroxyisonicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.