missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dihydroxybenzoic acid, MedChemExpress

Product Code. 16452718
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452718 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452718 Supplier Medchem Express Supplier No. HYY0801
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 2,6-Dihydroxybenzoic acid is a secondary metabolite of salicylic acid which has been hydrolyzed by liver enzymes during phase I metabolism.

2,6-Dihydroxybenzoic acid is a secondary metabolite of salicylic acid which has been hydrolyzed by liver enzymes during phase I metabolism.

Specifications

Chemical Name or Material 2,6-Dihydroxybenzoic acid
CAS 303-07-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C7H6O4
Quantity 5 g
Solubility Information DMSO : 100 mg/mL (648.85 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (108.16 mM; Need ultrasonic)
SMILES O=C(O)C1=C(O)C=CC=C1O
Molecular Weight (g/mol) 154.12
Formula Weight 154.12
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.