missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Dihydroxyacetophenone, Ultrapure MALDI Matrix

Product Code. 15458769
Change view
Click to view available options
Quantity:
5 x 10 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458769 5 x 10 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15458769 Supplier Alfa Aesar™ Supplier No. J67625.!K
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use as a matrix in MALDI mass spectrometry analysis

TRUSTED_SUSTAINABILITY

Specifications

CAS 699-83-2
Melting Point 156°C to 159°C
Color Brown to Yellow
Molecular Formula C8H8O3
MDL Number MFCD00002270
Quantity 5 x 10 mg
Beilstein 1366061
Synonym 2',6'-dihydroxyacetophenone, 1-2,6-dihydroxyphenyl ethanone, 2,6-dihydroxyacetophenone, 2-acetylresorcinol, resorcinol, 2-acetyl, 1-2,6-dihydroxyphenyl ethan-1-one, 2,6-dihydroxy acetophenone, gamma-resacetophenone, ethanone, 1-2,6-dihydroxyphenyl, unii-88bo51g3y2
InChI Key YPTJKHVBDCRKNF-UHFFFAOYSA-N
SMILES CC(=O)C1=C(C=CC=C1O)O
IUPAC Name 1-(2,6-dihydroxyphenyl)ethanone
Molecular Weight (g/mol) 152.149
PubChem CID 69687
Formula Weight 152.15
Physical Form Powder
Chemical Name or Material 2,6-Dihydroxyacetophenone, Ultrapure MALDI Matrix
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.