missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dihydroxyacetophenone, 97%

Product Code. 10264520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10264520 50 g Glass bottle 50g
10764411 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10264520 Supplier Thermo Scientific Acros Supplier No. 114820500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 699-83-2 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699-83-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002270
InChI Key YPTJKHVBDCRKNF-UHFFFAOYSA-N
Synonym 2',6'-dihydroxyacetophenone, 1-2,6-dihydroxyphenyl ethanone, 2,6-dihydroxyacetophenone, 2-acetylresorcinol, resorcinol, 2-acetyl, 1-2,6-dihydroxyphenyl ethan-1-one, 2,6-dihydroxy acetophenone, gamma-resacetophenone, ethanone, 1-2,6-dihydroxyphenyl, unii-88bo51g3y2
PubChem CID 69687
IUPAC Name 1-(2,6-dihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC=C1O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 156.0°C to 159.0°C
Color Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (HO)2C6H3COCH3
Quantity 50 g
Solubility Information Solubility in water: sparingly soluble. Other solubilities: soluble in ethanol,acetone,pyridine and dmf
Formula Weight 152.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2', 6'-Dihydroxyacetophenone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2',6'-Dihydroxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.