missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dihydroxyacetophenone, 98+%

Product Code. 11477576
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477576 100 g 100g
11457576 5 g 5g
11467576 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11477576 Supplier Thermo Scientific Alfa Aesar Supplier No. A14627.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 699-83-2 | C8H8O3 | 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699-83-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002270
InChI Key YPTJKHVBDCRKNF-UHFFFAOYSA-N
Synonym 2',6'-dihydroxyacetophenone, 1-2,6-dihydroxyphenyl ethanone, 2,6-dihydroxyacetophenone, 2-acetylresorcinol, resorcinol, 2-acetyl, 1-2,6-dihydroxyphenyl ethan-1-one, 2,6-dihydroxy acetophenone, gamma-resacetophenone, ethanone, 1-2,6-dihydroxyphenyl, unii-88bo51g3y2
PubChem CID 69687
IUPAC Name 1-(2,6-dihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC=C1O)O

Specifications

Melting Point 156°C to 159°C
Quantity 100 g
Beilstein 1366061
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 2',6'-Dihydroxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.