missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoropyridine, 99%

Product Code. 10361524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10361524 25 g Glass bottle 25g
10093660 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10361524 Supplier Thermo Scientific Acros Supplier No. 306660250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1513-65-1 | C5H3F2N | 115.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1513-65-1
Molecular Formula C5H3F2N
Molecular Weight (g/mol) 115.08
MDL Number MFCD00006227
InChI Key MBTGBRYMJKYYOE-UHFFFAOYSA-N
Synonym pyridine, 2,6-difluoro, 2,6-difluorpyridin, ccris 1720, 2,6-difluoro-pyridine, 2,6-difluoropyridin, pubchem9241, 2,6 difluoropyridine, 2, 6-difluoropyridine, 2,6 difluoro pyridine, 2,6-di-fluoropyridine
PubChem CID 73934
IUPAC Name 2,6-difluoropyridine
SMILES C1=CC(=NC(=C1)F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Purple-Red
Density 1.2600g/mL
Boiling Point 124.5°C (743.0 mmHg)
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4360 to 1.4380
Quantity 25 g
Specific Gravity 1.26
Formula Weight 115.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 6-Difluoropyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.