missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoropyridine, 98+%

Product Code. 11442937
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442937 100 g 100g
11422937 5 g 5g
11432937 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442937 Supplier Thermo Scientific Alfa Aesar Supplier No. A15522.22
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 1513-65-1 | C5H3F2N | 115.083 g/mol

2,6-Difluoropyridine has been used in the preparation of poly(pyridine ether)s via polycondensation with silylated 1,1,1-tris(4-hydroxyphenyl)ethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluoropyridine has been used in the preparation of poly(pyridine ether)s via polycondensation with silylated 1,1,1-tris(4-hydroxyphenyl)ethane.

Solubility
Not miscible or difficult to mix.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1513-65-1
Molecular Formula C5H3F2N
Molecular Weight (g/mol) 115.083
MDL Number MFCD00006227
InChI Key MBTGBRYMJKYYOE-UHFFFAOYSA-N
Synonym pyridine, 2,6-difluoro, 2,6-difluorpyridin, ccris 1720, 2,6-difluoro-pyridine, 2,6-difluoropyridin, pubchem9241, 2,6 difluoropyridine, 2, 6-difluoropyridine, 2,6 difluoro pyridine, 2,6-di-fluoropyridine
PubChem CID 73934
IUPAC Name 2,6-difluoropyridine
SMILES C1=CC(=NC(=C1)F)F

Specifications

Density 1.268
Boiling Point 123°C to 124°C
Flash Point 33°C (91°F)
Odor Characteristic
Refractive Index 1.437
Quantity 100 g
UN Number UN1993
Beilstein 1422549
Solubility Information Not miscible or difficult to mix.
Formula Weight 115.08
Percent Purity ≥98%
Chemical Name or Material 2,6-Difluoropyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.