missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoropyridine-3-boronic acid, 95%, Thermo Scientific™

Product Code. 11481760
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11481760

Brand: Thermo Scientific Alfa Aesar H27509.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136466-94-9
Molecular Formula C5H4BF2NO2
Molecular Weight (g/mol) 158.899
MDL Number MFCD03788245
InChI Key LCCZTROJRJFXNV-UHFFFAOYSA-N
Synonym 2,6-difluoropyridine-3-boronic acid, 2,6-difluoropyridin-3-yl boronic acid, 2,6-difluoro-3-pyridineboronic acid, 2,6-difluoropyridin-3-yl-3-boronic acid, 2,6-difluoropyridine-3-boronicacid, 2,6-difluoropyridin-3-ylboronicacid, 2,6-difluoro-3-pyridyl boronic acid, 2,6-difluoro-3-pyridylboronic acid, boronic acid, 2,6-difluoro-3-pyridinyl, 2,6-difluoro-3-pyridinyl boronic acid
PubChem CID 2762713
IUPAC Name (2,6-difluoropyridin-3-yl)boronic acid
SMILES B(C1=C(N=C(C=C1)F)F)(O)O

Specifications

Quantity 1 g
Formula Weight 158.9
Percent Purity 95%
Chemical Name or Material 2,6-Difluoropyridine-3-boronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.