missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorophenylboronic acid, 97%, ACROS Organics™

Product Code. 10487962 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10487962 1 g Glass bottle 1g
10212802 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10487962 Supplier Thermo Scientific Acros Supplier No. 359150010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 162101-25-9
Molecular Formula C6H5BF2O2
Molecular Weight (g/mol) 157.911
InChI Key DBZAICSEFBVFHL-UHFFFAOYSA-N
Synonym 2,6-difluorobenzeneboronic acid, 2,6-difluorophenyl boronic acid, 2,6-difluorophenyl boranediol, boronic acid, 2,6-difluorophenyl, 2,6-difluorphenylboronic acid, 2,6-difluoro phenylboric acid, boronic acid, b-2,6-difluorophenyl, 2,6-difluorophenylboricacid, pubchem1816, acmc-1celt
PubChem CID 2734336
IUPAC Name (2,6-difluorophenyl)boronic acid
SMILES B(C1=C(C=CC=C1F)F)(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 157.91
Percent Purity ≥96.5%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Difluorophenylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.