missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorophenyl isothiocyanate, 97%

Product Code. 11342707
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11342707

Brand: Thermo Scientific Alfa Aesar L11903.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 207974-17-2 | C7H3F2NS | 171.165 g/mol

2,6-Difluorophenyl isothiocyanate is used as chemical, organic and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluorophenyl isothiocyanate is used as chemical, organic and pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to moisture. Incompatible with amines, alcohols and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207974-17-2
Molecular Formula C7H3F2NS
Molecular Weight (g/mol) 171.165
MDL Number MFCD00041047
InChI Key DBSXNGIBAKYMSS-UHFFFAOYSA-N
PubChem CID 4031601
IUPAC Name 1,3-difluoro-2-isothiocyanatobenzene
SMILES C1=CC(=C(C(=C1)F)N=C=S)F

Specifications

Density 1.325
Boiling Point 217°C
Flash Point 104°C (219°F)
Refractive Index 1.602
Quantity 5 g
UN Number UN2922
Beilstein 2090586
Sensitivity Moisture sensitive
Synonym 2,6-difluorophenyl isothiocyanate, 2,6-difluorophenylisothiocyanate, benzene, 1,3-difluoro-2-isothiocyanato, 2,6-difluorobenzenisothiocyanate, pubchem5021, acmc-1caus, 2,6-difluorophenyl-isothiocyanate, 1,3-difluoro-2-isothiocyanato-benzene, benzene,1,3-difluoro-2-isothiocyanato
Solubility Information Hydrolyzes in water.
Formula Weight 171.17
Percent Purity 97%
Chemical Name or Material 2,6-Difluorophenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.