missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluorophenyl isothiocyanate, 97%
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
2,6-Difluorophenyl isothiocyanate is used as chemical, organic and pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Difluorophenyl isothiocyanate is used as chemical, organic and pharmaceutical intermediate.
Solubility
Hydrolyzes in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to moisture. Incompatible with amines, alcohols and oxidizing agents.
2,6-Difluorophenyl isothiocyanate is used as chemical, organic and pharmaceutical intermediate.
Solubility
Hydrolyzes in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to moisture. Incompatible with amines, alcohols and oxidizing agents.
Specifications
Specifications
| Density | 1.325 |
| Boiling Point | 217°C |
| Flash Point | 104°C (219°F) |
| Refractive Index | 1.602 |
| Quantity | 1 g |
| UN Number | UN2922 |
| Beilstein | 2090586 |
| Sensitivity | Moisture sensitive |
| Synonym | 2,6-difluorophenyl isothiocyanate, 2,6-difluorophenylisothiocyanate, benzene, 1,3-difluoro-2-isothiocyanato, 2,6-difluorobenzenisothiocyanate, pubchem5021, acmc-1caus, 2,6-difluorophenyl-isothiocyanate, 1,3-difluoro-2-isothiocyanato-benzene, benzene,1,3-difluoro-2-isothiocyanato |
| Solubility Information | Hydrolyzes in water. |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction