missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorophenyl isocyanate, 97%, ACROS Organics™

Catalog Number p-3757693
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
65295-69-4
C7H3F2NO
155.104
MFCD00001998
YXHDLKWTPVMIOH-UHFFFAOYSA-N
2,6-difluorophenyl isocyanate, 2,6-difluorophenylisocyanate, benzene, 1,3-difluoro-2-isocyanato, 2,6-difluorobenzenisocyanate, timtec-bb sbb006664, pubchem5023, acmc-1bcwu, 2,6 difluorophenylisocyanate, 2,6-difluoro phenyl isocyanate, 1-isocyanato-2,6-difluoro benzene
522162
1,3-difluoro-2-isocyanatobenzene
C1=CC(=C(C(=C1)F)N=C=O)F
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
65295-69-4
C7H3F2NO
155.104
MFCD00001998
YXHDLKWTPVMIOH-UHFFFAOYSA-N
2,6-difluorophenyl isocyanate, 2,6-difluorophenylisocyanate, benzene, 1,3-difluoro-2-isocyanato, 2,6-difluorobenzenisocyanate, timtec-bb sbb006664, pubchem5023, acmc-1bcwu, 2,6 difluorophenylisocyanate, 2,6-difluoro phenyl isocyanate, 1-isocyanato-2,6-difluoro benzene
522162
1,3-difluoro-2-isocyanatobenzene
C1=CC(=C(C(=C1)F)N=C=O)F
CAS Min % 96.0
CAS Max % 100.0
Melting Point 22.0°C to 25.0°C
Color White to Yellow
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula F2C6H3NCO
Refractive Index 1.4945 to 1.4965
Quantity 1 g
Formula Weight 155.1
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2, 6-Difluorophenyl isocyanate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'