missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzyl chloride, TRC

Product Code. 30376924
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376924 2.5 g 2.5g
30369062 1 g -
30370871 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376924 Supplier Toronto Research Chemicals Supplier No. TRCD4514582.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697-73-4
Molecular Formula C7 H5 Cl F2
Molecular Weight (g/mol) 162.56
Synonym 2-(chloromethyl)-1,3-difluoro-benzene
IUPAC Name 2-(chloromethyl)-1,3-difluoro-benzene
SMILES Fc1cccc(F)c1CCl

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H5ClF2/c8-4-5-6(9)2-1-3-7(5)10/h1-3H,4H2
Formula Weight 162.0048
Chemical Name or Material 2,6-Difluorobenzyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.