missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11494047
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494047 25 g 25g
11404060 1 g 1g
11484047 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494047 Supplier Thermo Scientific Alfa Aesar Supplier No. A19472.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2,6-Difluorobenzyl chloride is used as rufinamide intermediate. It is an important raw material and intermediate used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluorobenzyl chloride is used as rufinamide intermediate. It is an important raw material and intermediate used in organic synthesis.

Solubility
Soluble in acetone, DMSO and DMF. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697-73-4
Molecular Formula C7H5ClF2
Molecular Weight (g/mol) 162.564
MDL Number MFCD00191346
InChI Key MJXRENZUAQXZGJ-UHFFFAOYSA-N
Synonym 2,6-difluorobenzyl chloride, 2-chloromethyl-1,3-difluorobenzene, 2,6-difluorobenzylchloride, benzene, 2-chloromethyl-1,3-difluoro, alpha-chloro-2,6-difluorotoluene, 2, 6-difluorobenzyl chloride, 1-chloromethyl-2,6-difluorobenzene, 2-chlormethyl-1,3-difluorbenzol, pubchem2178
PubChem CID 581570
IUPAC Name 2-(chloromethyl)-1,3-difluorobenzene
SMILES C1=CC(=C(C(=C1)F)CCl)F

Specifications

Melting Point 32°C to 35°C
Boiling Point 172°C to 173°C (640 mmHg)
Quantity 25 g
UN Number UN1760
Beilstein 243793
Formula Weight 162.57
Percent Purity 98%
Chemical Name or Material 2,6-Difluorobenzyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.