missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzyl bromide, 96%

Product Code. p-7023649
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11468017

Brand: Thermo Scientific Alfa Aesar A17434.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85118-00-9 | C7H5BrF2 | 207.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85118-00-9
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.02
MDL Number MFCD00000329
InChI Key LSXJPJGBWSZHTM-UHFFFAOYSA-N
Synonym 2,6-difluorobenzyl bromide, 2-bromomethyl-1,3-difluorobenzene, 2,6-difluorobenzylbromide, alpha-bromo-2,6-difluorotoluene, benzene, 2-bromomethyl-1,3-difluoro, timtec-bb sbb006564, 2-bromomethyl-1,3-difluoro-benzene, fr cf b1e, pubchem4915, acmc-209q4j
PubChem CID 581435
IUPAC Name 2-(bromomethyl)-1,3-difluorobenzene
SMILES FC1=CC=CC(F)=C1CBr

Specifications

Melting Point 49°C to 53°C
Density 1.6
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN1759
Beilstein 2083943
Formula Weight 207.02
Percent Purity ≥97%
Chemical Name or Material 2,6-Difluorobenzyl bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.