missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzoyl chloride, 99%, ACROS Organics™

Product Code. 10521282 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10521282 5mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10521282 Supplier Thermo Scientific Acros Supplier No. 224510050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

CAS 18063-02-0
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C7H3ClF2O
Linear Formula F2C6H3COCl
Refractive Index 1.4988 to 1.5008 (20°C, 589nm)
MDL Number MFCD00000659
Quantity 5 mL
Synonym 2,6-difluorobenzoylchloride, benzoyl chloride, 2,6-difluoro, 2,6-difluoro-benzoyl chloride, acmc-1ca8w, 2,6-diflurobenzoyl chloride, 2 6-difluorobenzoyl chloride, 2,6-diflourobenzoyl chloride, 2,6-difluoro benzoylchloride, 2.6-difluorobenzoyl chloride, 2,6-difluro-benzoyl chloride
InChI Key QRHUZEVERIHEPT-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)F)C(=O)Cl)F
IUPAC Name 2,6-difluorobenzoyl chloride
Molecular Weight (g/mol) 176.547
PubChem CID 87438
Formula Weight 176.55
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 2, 6-Difluorobenzoyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,6-Difluorobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.