missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluorobenzonitrile, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2,6-Difluorobenzonitrile |
| CAS | 1897-52-5 |
| Molecular Formula | C7H3F2N |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C7H3F2N/c8-6-2-1-3-7(9)5(6)4-10/h1-3H |
| SMILES | Fc1cccc(F)c1C#N |
| IUPAC Name | 2,6-difluorobenzonitrile |
| Molecular Weight (g/mol) | 139.1 |
| Formula Weight | 139.0234 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction