missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzonitrile, 98%

Product Code. 11478673
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478673 250 g 250g
11458673 10 g 10g
11468673 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478673 Supplier Thermo Scientific Alfa Aesar Supplier No. A10454.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1897-52-5 | C7H3F2N | 139.105 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1897-52-5
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.105
MDL Number MFCD00001778
InChI Key BNBRIFIJRKJGEI-UHFFFAOYSA-N
Synonym 2,6-difluorobenznitrile, benzonitrile, 2,6-difluoro, 2,6-difluoro-benzonitrile, 2,6-dfbn, unii-1j58z42a2e, 2,6-difluoro benzonitrile, 2,6-difluorobenzenecarbonitrile, 2,6-difluorophenylcyanide, ncr bf ff, 6-difluorobenzonitrile
PubChem CID 74695
IUPAC Name 2,6-difluorobenzonitrile
SMILES C1=CC(=C(C(=C1)F)C#N)F

Specifications

Melting Point 28°C to 32°C
Density 1.246
Boiling Point 197°C to 198°C
Flash Point 80°C (176°F)
Odor Almond-like
Refractive Index 1.4875
Quantity 250 g
UN Number UN3439
Beilstein 2045292
Formula Weight 139.11
Percent Purity 98%
Chemical Name or Material 2,6-Difluorobenzonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.