missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzeneboronic acid, 98%

Code produit. 11459837
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
missing translation for 'unitSize'
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantity unitSize
11459837 1 g 1g
11469837 5 g 5g
11479837 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 11459837 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' B22805.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 162101-25-9 | C6H5BF2O2 | 157.911 g/mol

2,6-Difluorobenzeneboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluorobenzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 162101-25-9
Molecular Formula C6H5BF2O2
Molecular Weight (g/mol) 157.911
MDL Number MFCD00792436
InChI Key DBZAICSEFBVFHL-UHFFFAOYSA-N
Synonym 2,6-difluorobenzeneboronic acid, 2,6-difluorophenyl boronic acid, 2,6-difluorophenyl boranediol, boronic acid, 2,6-difluorophenyl, 2,6-difluorphenylboronic acid, 2,6-difluoro phenylboric acid, boronic acid, b-2,6-difluorophenyl, 2,6-difluorophenylboricacid, pubchem1816, acmc-1celt
PubChem CID 2734336
IUPAC Name (2,6-difluorophenyl)boronic acid
SMILES B(C1=C(C=CC=C1F)F)(O)O

Spécification

Melting Point ∼140°C (decomposition)
Quantity 1 g
Beilstein 8545931
Solubility Information Soluble in methanol.
Formula Weight 157.91
Percent Purity 98%
Chemical Name or Material 2,6-Difluorobenzeneboronic acid

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.