missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzamide, TRC

Product Code. 30132796
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132796 5 g 5g
30132428 10 g 10g
30008812 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132796 Supplier Toronto Research Chemicals Supplier No. TRCD4518435G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18063-03-1
Molecular Formula C7 H5 F2 N O
Molecular Weight (g/mol) 157.12
Synonym Benzamide, 2,6-difluoro-, 2,6-Difluorobenzamide, Lufenuron Imp. A (EP)
IUPAC Name 2,6-difluorobenzamide
SMILES NC(=O)c1c(F)cccc1F

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H5F2NO/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H2,10,11)
Formula Weight 157.0339
Chemical Name or Material 2,6-Difluorobenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.