missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzaldehyde, 98%

Product Code. 10219753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10219753 25 g Glass bottle 25g
10480341 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10219753 Supplier Thermo Scientific Acros Supplier No. 311460250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 437-81-0 | C7H4F2O | 142.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 437-81-0
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.11
MDL Number MFCD00010293
InChI Key SOWRUJSGHKNOKN-UHFFFAOYSA-N
Synonym 2,6-difluoro benzaldehyde, 2,6-dfad, benzaldehyde, 2,6-difluoro, 2,6-difluorobenzaledehyde, pubchem1438, 2,6-difluorbenzaldehyde, 2,6 difluorobenzaldehyde, acmc-1adi4, 2, 6-difluorobenzaldehyde, 2,6-difluoro-benzaldehyde
PubChem CID 136284
IUPAC Name 2,6-difluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)F)C=O)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 15°C to 17°C
Color Yellow
Density 1.3100g/mL
Boiling Point 82°C to 84°C (15.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3COH
Refractive Index 1.5060 to 1.509
Quantity 25 g
Specific Gravity 1.31
Formula Weight 142.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 6-Difluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.