missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzaldehyde, 97%

Product Code. 11499246
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499246 25 g 25g
11489246 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499246 Supplier Thermo Scientific Alfa Aesar Supplier No. A18099.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 437-81-0 | C7H4F2O | 142.105 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 437-81-0
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.105
MDL Number MFCD00010293
InChI Key SOWRUJSGHKNOKN-UHFFFAOYSA-N
Synonym 2,6-difluoro benzaldehyde, 2,6-dfad, benzaldehyde, 2,6-difluoro, 2,6-difluorobenzaledehyde, pubchem1438, 2,6-difluorbenzaldehyde, 2,6 difluorobenzaldehyde, acmc-1adi4, 2, 6-difluorobenzaldehyde, 2,6-difluoro-benzaldehyde
PubChem CID 136284
IUPAC Name 2,6-difluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)F)C=O)F

Specifications

Melting Point 15°C to 17°C
Density 1.317
Boiling Point 85°C to 86°C (20 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.507
Quantity 25 g
Beilstein 1935273
Sensitivity Air Sensitive
Formula Weight 142.11
Percent Purity 97%
Chemical Name or Material 2,6-Difluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.