missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoroaniline, 98%

Product Code. 11479933
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479933 100 g 100g
11459933 5 g 5g
11469933 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479933 Supplier Thermo Scientific Alfa Aesar Supplier No. A13020.22
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 5509-65-9 | C6H5F2N | 129.11 g/mol

2,6-Difluoroaniline is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluoroaniline is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to acid chlorides, acid anhydrides, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5509-65-9
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007655
InChI Key ODUZJBKKYBQIBX-UHFFFAOYSA-N
Synonym benzenamine, 2,6-difluoro, 2,6-difluoro-phenylamine, 2,6-difluoro aniline, 2,6-difluorobenzenamine, 2,6-difluoranilin, 2,6 difluoroaniline, 2,6-diflouroaniline, pubchem4034, 2,6-difuluoroaniline
PubChem CID 79647
IUPAC Name 2,6-difluoroaniline
SMILES NC1=C(F)C=CC=C1F

Specifications

Density 1.275
Boiling Point 152°C to 154°C
Flash Point 51°C (124°F)
Odor Characteristic
Refractive Index 1.508
Quantity 100 g
UN Number UN1992
Beilstein 2802697
Solubility Information Slightly soluble in water.
Formula Weight 129.11
Percent Purity 98%
Chemical Name or Material 2,6-Difluoroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.