missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Difluoroacetophenone, 98%

Product Code. 11429562
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429562 25 g 25g
11409562 1 g 1g
11419562 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429562 Supplier Thermo Scientific Alfa Aesar Supplier No. A10220.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13670-99-0 | C8H6F2O | 156.13 g/mol

2?,6?-Difluoroacetophenone was used in the synthesis of 2-amino-4-alkyl- and 2-amino-4-arylquinazolines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,6′-Difluoroacetophenone was used in the synthesis of 2-amino-4-alkyl- and 2-amino-4-arylquinazolines.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13670-99-0
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.13
MDL Number MFCD00000328
InChI Key VGIIILXIQLXVLC-UHFFFAOYSA-N
Synonym 2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054
PubChem CID 83643
SMILES CC(=O)C1=C(F)C=CC=C1F

Specifications

Density 1.195
Boiling Point 76°C to 79°C (15 mmHg)
Flash Point 76°C (168°F)
Refractive Index 1.481
Quantity 25 g
Beilstein 2046068
Solubility Information Slightly soluble in water.
IUPAC Name 1-(2,6-difluorophenyl)ethanone
Formula Weight 156.13
Percent Purity 98%
Chemical Name or Material 2',6'-Difluoroacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.