Learn More
2,6-Difluoro-3-nitrotoluene, 98%, Thermo Scientific™
£19.10 - £171.00
Chemical Identifiers
CAS | 79562-49-5 |
---|---|
Molecular Formula | C7H5F2NO2 |
Molecular Weight (g/mol) | 173.119 |
MDL Number | MFCD11519030 |
InChI Key | AVWNNVJXXMKAPB-UHFFFAOYSA-N |
Synonym | 2,6-difluoro-3-nitrotoluene, benzene, 1,3-difluoro-2-methyl-4-nitro, 2,4-difluoro-3-methylnitrobenzene, pubchem10410, pubchem15439, 1,3-difluoro-2-methyl-4-nitro-benzene, 1,3-bis fluoranyl-2-methyl-4-nitro-benzene |
PubChem CID | 12795486 |
IUPAC Name | 1,3-difluoro-2-methyl-4-nitrobenzene |
SMILES | CC1=C(C=CC(=C1F)[N+](=O)[O-])F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15480119
|
Thermo Scientific Alfa Aesar
H63431.MD |
250 mg |
£19.10
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15460119
|
Thermo Scientific Alfa Aesar
H63431.03 |
1 g | |||||||
15470119
|
Thermo Scientific Alfa Aesar
H63431.06 |
5 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2,6-Difluoro-3-nitrotoluene used as a intermediate in organic synthesis, pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
79562-49-5 | |
173.119 | |
AVWNNVJXXMKAPB-UHFFFAOYSA-N | |
12795486 | |
CC1=C(C=CC(=C1F)[N+](=O)[O-])F |
C7H5F2NO2 | |
MFCD11519030 | |
2,6-difluoro-3-nitrotoluene, benzene, 1,3-difluoro-2-methyl-4-nitro, 2,4-difluoro-3-methylnitrobenzene, pubchem10410, pubchem15439, 1,3-difluoro-2-methyl-4-nitro-benzene, 1,3-bis fluoranyl-2-methyl-4-nitro-benzene | |
1,3-difluoro-2-methyl-4-nitrobenzene |
Specifications
79562-49-5 | |
MFCD11519030 | |
2,6-difluoro-3-nitrotoluene, benzene, 1,3-difluoro-2-methyl-4-nitro, 2,4-difluoro-3-methylnitrobenzene, pubchem10410, pubchem15439, 1,3-difluoro-2-methyl-4-nitro-benzene, 1,3-bis fluoranyl-2-methyl-4-nitro-benzene | |
AVWNNVJXXMKAPB-UHFFFAOYSA-N | |
1,3-difluoro-2-methyl-4-nitrobenzene | |
12795486 | |
98% | |
2,6-Difluoro-3-nitrotoluene |
C7H5F2NO2 | |
UN2811 | |
Slightly soluble in water. | |
CC1=C(C=CC(=C1F)[N+](=O)[O-])F | |
173.119 | |
173.12 | |
Solid |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures