missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-3-methylaniline, 97%, Thermo Scientific™

Product Code. 11372559
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372559 1 g 1g
11382559 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372559 Supplier Thermo Scientific Alfa Aesar Supplier No. H26656.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,6-Difluoro-3-methylaniline is used as a solvent is convenient as the pyridine can serve both as the solvent and the catalyst in the transformation.

Solubility
Soluble in organic solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144851-63-8
Molecular Formula C7H7F2N
Molecular Weight (g/mol) 143.14
MDL Number MFCD06660182
InChI Key ZMNJSSZHDRBGNN-UHFFFAOYSA-N
Synonym benzenamine, 2,6-difluoro-3-methyl, 2,6-difluoro-m-toluidine, 3-amino-2,4-difluorotoluene, 2,6-difluoro-3-methylphenylamine
PubChem CID 15017536
IUPAC Name 2,6-difluoro-3-methylaniline
SMILES CC1=C(F)C(N)=C(F)C=C1

Specifications

Quantity 1 g
UN Number UN2810
Solubility Information Soluble in organic solvents.
Formula Weight 143.14
Percent Purity 97%
Chemical Name or Material 2,6-Difluoro-3-methylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.