missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorotoluene, 99%

Product Code. 11425927
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425927 100 g 100g
11415927 25 g 25g
11435927 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425927 Supplier Thermo Scientific Alfa Aesar Supplier No. B24463.22
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 118-69-4 | C7H6Cl2 | 161.025 g/mol

2,6-Dichlorotoluene is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichlorotoluene is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-69-4
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000576
InChI Key DMEDNTFWIHCBRK-UHFFFAOYSA-N
Synonym 2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, toluene, 2,6-dichloro, unii-rvm09757ww, 1,3-dichloro-2-methyl-benzene, toluene,6-dichloro, pubchem3701, 2,6-dichloro-toluene, benzene,dichloromethyl, 2,6-dichloro toluene
PubChem CID 8368
IUPAC Name 1,3-dichloro-2-methylbenzene
SMILES CC1=C(C=CC=C1Cl)Cl

Specifications

Melting Point 2°C
Density 1.254
Boiling Point 196°C to 203°C
Flash Point 82°C (179°F)
Refractive Index 1.551
Quantity 100 g
UN Number UN3082
Beilstein 1907194
Solubility Information Soluble in water.
Formula Weight 161.03
Percent Purity 99%
Chemical Name or Material 2,6-Dichlorotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.