missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorotoluene, 99+%

Product Code. 10142380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10142380 500 mL Glass bottle 500mL
10590371 100 mL Glass bottle 100mL
10541184 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10142380 Supplier Thermo Scientific Acros Supplier No. 113795000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118-69-4 | C7H6Cl2 | 161.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-69-4
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.03
MDL Number MFCD00000576
InChI Key DMEDNTFWIHCBRK-UHFFFAOYSA-N
Synonym 2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, toluene, 2,6-dichloro, unii-rvm09757ww, 1,3-dichloro-2-methyl-benzene, toluene,6-dichloro, pubchem3701, 2,6-dichloro-toluene, benzene,dichloromethyl, 2,6-dichloro toluene
PubChem CID 8368
IUPAC Name 1,3-dichloro-2-methylbenzene
SMILES CC1=C(C=CC=C1Cl)Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 2°C
Color Colorless to Yellow
Density 1.2500g/mL
Boiling Point 196°C to 203°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3Cl2
Refractive Index 1.5500 to 1.552
Quantity 500 mL
Beilstein 05, 296
Specific Gravity 1.25
Solubility Information Solubility in water: 24mg/l (20°C). Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 161.03
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2, 6-Dichlorotoluene, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,6-Dichlorotoluene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.