missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorothiophenol, 97%

Product Code. 11485263
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485263 25 g 25g
11475263 5 g 5g
11495263 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485263 Supplier Thermo Scientific Alfa Aesar Supplier No. A12854.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24966-39-0 | C6H3Cl2S | 178.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24966-39-0
Molecular Formula C6H3Cl2S
Molecular Weight (g/mol) 178.05
MDL Number MFCD00004833
InChI Key JBISHCXLCGVPGW-UHFFFAOYSA-M
Synonym 2,6-dichlorothiophenol, 2,6-dichlorobenzene-1-thiol, 2,6-dichloro thiophenol, benzenethiol, 2,6-dichloro, pubchem6802, 2,6-dichloro-benzenethiol, acmc-209gg5, benzenethiol,2,6-dichloro
PubChem CID 90671
IUPAC Name 2,6-dichlorobenzenethiol
SMILES [S-]C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 46°C to 50°C
Boiling Point 101°C to 103°C (14 mmHg)
Flash Point 112°C (233°F)
Odor Unpleasant
Quantity 25 g
Beilstein 2082400
Sensitivity Air Sensitive
Formula Weight 179.07
Percent Purity 97%
Chemical Name or Material 2,6-Dichlorothiophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.