missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyridine N-oxide, 98%

Product Code. 15413937
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413937 1 g 1g
15423937 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15413937 Supplier Thermo Scientific Alfa Aesar Supplier No. H30427.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2587-00-0 | C5H3Cl2NO | 163.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2587-00-0
Molecular Formula C5H3Cl2NO
Molecular Weight (g/mol) 163.99
MDL Number MFCD00130246
InChI Key RFOMGVDPYLWLOC-UHFFFAOYSA-N
Synonym 2,6-dichloropyridine n-oxide, 2,6-dichloropyridine 1-oxide, 2,6-dichloropyridine-n-oxide, 2,6-dichloropyridin-1-ium-1-olate, pyridine, 2,6-dichloro-, 1-oxide, 2,6-dichloropyridine-1-oxide, 2,6-dichloropyridine n-o, ide, acmc-1cjmr, 2,6-dichloropyridin-1-ol
PubChem CID 817673
IUPAC Name 2,6-dichloro-1-oxidopyridin-1-ium
SMILES [O-][N+]1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 136°C to 140°C
Quantity 1 g
UN Number UN2811
Formula Weight 163.99
Percent Purity 98%
Chemical Name or Material 2,6-Dichloropyridine N-oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.