missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyridine, 98%

Product Code. 11420173
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420173 500 g 500g
11410173 100 g 100g
11430173 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420173 Supplier Thermo Scientific Alfa Aesar Supplier No. A11364.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2402-78-0 | C5H3Cl2N | 147.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2402-78-0
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006244
InChI Key FILKGCRCWDMBKA-UHFFFAOYSA-N
Synonym pyridine, 2,6-dichloro, ccris 1727, 2,6-dichlorpyridine, 2.6-dichloropyridine, pubchem2564, 2,6-dichloro pyridine, 2,6-dichloro-pyridine, acmc-1cbwr, 26dclpy, wln: t6nj bg fg
PubChem CID 16989
IUPAC Name 2,6-dichloropyridine
SMILES ClC1=CC=CC(Cl)=N1

Specifications

Melting Point 84°C to 88°C
Boiling Point 211°C
Odor Unpleasant
Quantity 500 g
UN Number UN2811
Beilstein 108664
Formula Weight 147.99
Percent Purity 98%
Chemical Name or Material 2,6-Dichloropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.