missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyridin-4-amine, 97%, Thermo Scientific™

Product Code. 10754203
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10754203

Brand: Thermo Scientific Maybridge RF04500DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 2587-02-2, 2588-04-7
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H5Cl2N2
Quantity 1 g
Synonym 4-amino-2,6-dichloropyridine, 2,6-dichloro-4-aminopyridine, 2,6-dichloro-pyridin-4-ylamine, 4-pyridinamine, 2,6-dichloro, 2,6-dichloro-4-pyridylamine, 2,6-dichloropyridin-4-ylamine, 2,6-dichloro-4-amino pyridine, 2,6-dichloro-4-pyridinamine, pubchem5530, acmc-1cot2
InChI Key WAEZOSSWRXDWAX-UHFFFAOYSA-O
SMILES C1=C(C=C(N=C1Cl)Cl)N
IUPAC Name 2,6-dichloropyridin-4-amine
Molecular Weight (g/mol) 164.01
PubChem CID 137652
Formula Weight 163.01
Percent Purity 97%
Chemical Name or Material 2, 6-Dichloropyridin-4-amine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.